Assay Details
| Gene Symbol: LOC4337574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784553.2 | XP_015640039.1 | 10 - 11 | 2267 | 119 | |
| GenBank mRNA | AK099809.1 | - | 3 - 4 | 622 | 119 | |
| AK120821.1 | - | 7 - 8 | 1463 | 119 | ||
More Information
Set Membership: |
|

LOC4337574
Genomic Map