Assay Details
| Gene Symbol: LOC4338480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784840.1 | XP_015640326.1 | 10 - 11 | 1435 | 76 | |
| XM_015784841.2 | XP_015640327.1 | 8 - 9 | 1017 | 76 | ||
| GenBank mRNA | AK073748.1 | - | 10 - 11 | 1430 | 76 | |
| AK102482.1 | - | 10 - 11 | 1429 | 76 | ||
More Information
Set Membership: |
|

LOC4338480
Genomic Map