Assay Details
| Gene Symbol: LOC4338480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784840.1 | XP_015640326.1 | 16 - 17 | 2181 | 77 | |
| XM_015784841.2 | XP_015640327.1 | 14 - 15 | 1763 | 77 | ||
| GenBank mRNA | AK073748.1 | - | 16 - 17 | 2176 | 77 | |
| AK102482.1 | - | 16 - 17 | 2175 | 77 | ||
More Information
Set Membership: |
|

LOC4338480
Genomic Map