Assay Details
| Gene Symbol: LOC4338480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784840.1 | XP_015640326.1 | 1 - 2 | 282 | 77 | |
| GenBank mRNA | AK073748.1 | - | 1 - 2 | 277 | 77 | |
| AK102482.1 | - | 1 - 2 | 276 | 77 | ||
More Information
Set Membership: |
|

LOC4338480
Genomic Map