Assay Details
| Gene Symbol: LOC4338480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784840.1 | XP_015640326.1 | 2 - 3 | 435 | 121 | |
| GenBank mRNA | AK073748.1 | - | 2 - 3 | 430 | 121 | |
| AK102482.1 | - | 2 - 3 | 429 | 121 | ||
More Information
Set Membership: |
|

LOC4338480
Genomic Map