Assay Details
Gene Symbol: LOC4339576 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015781987.2 | XP_015637473.1 | 3 - 4 | 881 | 73 | |
GenBank mRNA | AK069566.1 | - | 3 - 4 | 749 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
