Assay Details
Gene Symbol: LOC4339576 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015781987.2 | XP_015637473.1 | 4 - 5 | 952 | 122 | |
GenBank mRNA | AK069566.1 | - | 4 - 5 | 820 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
