Assay Details
Gene Symbol: LOC4339576 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015781987.2 | XP_015637473.1 | 5 - 6 | 1136 | 87 | |
GenBank mRNA | AK069566.1 | - | 5 - 6 | 1004 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
