Assay Details
| Gene Symbol: LOC4340558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788648.2 | XP_015644134.1 | 4 - 5 | 816 | 58 | |
| GenBank mRNA | AB253627.1 | - | 4 - 5 | 647 | 58 | |
| AK072966.1 | - | 4 - 5 | 812 | 58 | ||
| AK104037.1 | - | 4 - 5 | 802 | 58 | ||
| AK112022.1 | - | 4 - 5 | 813 | 58 | ||
More Information
Set Membership: |
|

LOC4340558
Genomic Map