Assay Details
| Gene Symbol: LOC4340682 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787058.2 | XP_015642544.1 | 3 - 4 | 562 | 58 | |
| GenBank mRNA | AK108686.1 | - | 3 - 4 | 568 | 58 | |
More Information
Set Membership: |
|

LOC4340682
Genomic Map