Assay Details
Gene Symbol: LOC4340746 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015786313.2 | XP_015641799.1 | 1 - 2 | 1019 | 60 | |
GenBank mRNA | AB041838.1 | - | 1 - 2 | 984 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
