Assay Details
| Gene Symbol: LOC4341801 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788562.2 | XP_015644048.1 | 19 - 20 | 2483 | 95 | |
| GenBank mRNA | AK287475.1 | - | 19 - 20 | 2469 | 95 | |
More Information
Set Membership: |
|

LOC4341801
Genomic Map