Assay Details
| Gene Symbol: LOC4342680 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791038.1 | XP_015646524.1 | 12 - 13 | 1595 | 94 | |
| GenBank mRNA | AF054629.2 | - | 8 - 9 | 946 | 94 | |
| AK062079.1 | - | 6 - 7 | 728 | 94 | ||
| AK065213.1 | - | 12 - 13 | 1564 | 94 | ||
| AK066223.1 | - | 9 - 10 | 983 | 94 | ||
More Information
Set Membership: |
|

LOC4342680
Genomic Map