Assay Details
Gene Symbol: LOC4342680 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791038.1 | XP_015646524.1 | 5 - 6 | 747 | 74 | |
GenBank mRNA | AF054629.2 | - | 1 - 2 | 98 | 74 | |
AK065213.1 | - | 5 - 6 | 716 | 74 | ||
AK066223.1 | - | 2 - 3 | 135 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
