Assay Details
Gene Symbol: LOC4342680 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791038.1 | XP_015646524.1 | 6 - 7 | 852 | 64 | |
GenBank mRNA | AF054629.2 | - | 2 - 3 | 203 | 64 | |
AK066223.1 | - | 3 - 4 | 240 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
