Assay Details
Gene Symbol: LOC4342680 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791038.1 | XP_015646524.1 | 9 - 10 | 1231 | 143 | |
GenBank mRNA | AF054629.2 | - | 5 - 6 | 582 | 143 | |
AK062079.1 | - | 3 - 4 | 364 | 143 | ||
AK065213.1 | - | 9 - 10 | 1200 | 143 | ||
AK066223.1 | - | 6 - 7 | 619 | 143 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
