Assay Details
Gene Symbol: LOC4342729 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015790193.2 | XP_015645679.1 | 1 | 69 | 82 | |
GenBank mRNA | AK107674.1 | - | 1 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015790193.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
