Assay Details
Gene Symbol: LOC4342802 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015792161.1 | XP_015647647.1 | 2 - 3 | 434 | 99 | |
GenBank mRNA | AK068609.1 | - | 2 - 3 | 417 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
