Assay Details
Gene Symbol: LOC4342802 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015792161.1 | XP_015647647.1 | 7 - 8 | 852 | 104 | |
GenBank mRNA | AK068609.1 | - | 7 - 8 | 835 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
