Assay Details
Gene Symbol: LOC4342981 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791973.1 | XP_015647459.1 | 10 - 11 | 1501 | 67 | |
GenBank mRNA | AB110202.1 | - | 10 - 11 | 1047 | 67 | |
AK102570.1 | - | 6 - 7 | 1804 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
