Assay Details
| Gene Symbol: LOC4342981 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791973.1 | XP_015647459.1 | 1 - 2 | 568 | 70 | |
| GenBank mRNA | AB110202.1 | - | 1 - 2 | 114 | 70 | |
| AK073463.1 | - | 1 - 2 | 574 | 70 | ||
More Information
Set Membership: |
|

LOC4342981
Genomic Map