Assay Details
Gene Symbol: LOC4342981 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791973.1 | XP_015647459.1 | 2 - 3 | 695 | 57 | |
GenBank mRNA | AB110202.1 | - | 2 - 3 | 241 | 57 | |
AK073463.1 | - | 2 - 3 | 701 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
