Assay Details
| Gene Symbol: LOC4342981 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791973.1 | XP_015647459.1 | 4 - 5 | 875 | 101 | |
| GenBank mRNA | AB110202.1 | - | 4 - 5 | 421 | 101 | |
| AK073463.1 | - | 4 - 5 | 881 | 101 | ||
More Information
Set Membership: |
|

LOC4342981
Genomic Map