Assay Details
| Gene Symbol: LOC4342981 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791973.1 | XP_015647459.1 | 5 - 6 | 955 | 81 | |
| GenBank mRNA | AB110202.1 | - | 5 - 6 | 501 | 81 | |
| AK073463.1 | - | 5 - 6 | 961 | 81 | ||
| AK102570.1 | - | 1 - 2 | 1258 | 81 | ||
More Information
Set Membership: |
|

LOC4342981
Genomic Map