Assay Details
| Gene Symbol: LOC4342981 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791973.1 | XP_015647459.1 | 7 - 8 | 1165 | 105 | |
| GenBank mRNA | AB110202.1 | - | 7 - 8 | 711 | 105 | |
| AK073463.1 | - | 7 - 8 | 1171 | 105 | ||
| AK102570.1 | - | 3 - 4 | 1468 | 105 | ||
More Information
Set Membership: |
|

LOC4342981
Genomic Map