Assay Details
Gene Symbol: LOC4342981 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791973.1 | XP_015647459.1 | 8 - 9 | 1242 | 80 | |
GenBank mRNA | AB110202.1 | - | 8 - 9 | 788 | 80 | |
AK073463.1 | - | 8 - 9 | 1248 | 80 | ||
AK102570.1 | - | 4 - 5 | 1545 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
