Assay Details
Gene Symbol: LOC4342981 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791973.1 | XP_015647459.1 | 9 - 10 | 1335 | 151 | |
GenBank mRNA | AB110202.1 | - | 9 - 10 | 881 | 151 | |
AK073463.1 | - | 9 - 10 | 1341 | 151 | ||
AK102570.1 | - | 5 - 6 | 1638 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() |
