Assay Details
Gene Symbol: LOC4343014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015792247.2 | XP_015647733.1 | 1 | 1024 | 117 | |
GenBank mRNA | AK060411.1 | - | 1 | 833 | 117 | |
AK061478.1 | - | 1 | 953 | 117 | ||
AK104124.1 | - | 1 | 967 | 117 | ||
AK104173.1 | - | 1 | 827 | 117 | ||
AK120684.1 | - | 1 | 963 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
