Assay Details
| Gene Symbol: LOC4343256 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792381.2 | XP_015647867.1 | 1 - 2 | 460 | 79 | |
| GenBank mRNA | AK063998.1 | - | 1 - 2 | 465 | 79 | |
| AK068175.1 | - | 1 - 2 | 230 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015792381.2 transcripts.
Set Membership: |
|

LOC4343256
Genomic Map