Assay Details
| Gene Symbol: LOC4343256 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792381.2 | XP_015647867.1 | 2 - 3 | 536 | 59 | |
| GenBank mRNA | AK063998.1 | - | 2 - 3 | 541 | 59 | |
| AK068175.1 | - | 2 - 3 | 306 | 59 | ||
More Information
Set Membership: |
|

LOC4343256
Genomic Map