Assay Details
Gene Symbol: LOC4343412 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791089.2 | XP_015646575.1 | 14 - 15 | 3337 | 74 | |
GenBank mRNA | AK105311.1 | - | 2 - 3 | 348 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
