Assay Details
| Gene Symbol: LOC4343686 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789873.2 | XP_015645359.1 | 1 - 2 | 189 | 104 | |
| GenBank mRNA | AK073925.1 | - | 1 - 2 | 173 | 104 | |
More Information
Set Membership: |
|

LOC4343686
Genomic Map