Assay Details
Gene Symbol: LOC4343803 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791573.2 | XP_015647059.1 | 2 - 3 | 808 | 83 | |
XM_015791575.2 | XP_015647061.1 | 2 - 3 | 808 | 83 | ||
XM_015791576.2 | XP_015647062.1 | 2 - 3 | 808 | 83 | ||
GenBank mRNA | AK073674.1 | - | 2 - 3 | 662 | 83 | |
AK101648.1 | - | 2 - 3 | 727 | 83 | ||
AK122151.1 | - | 2 - 3 | 711 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
