Assay Details
| Gene Symbol: LOC4343829 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792187.2 | XP_015647673.1 | 2 - 3 | 452 | 62 | |
| XM_015792188.2 | XP_015647674.1 | 2 - 3 | 452 | 62 | ||
| GenBank mRNA | AK111892.1 | - | 2 - 3 | 429 | 62 | |
More Information
Set Membership: |
|

LOC4343829
Genomic Map