Assay Details
| Gene Symbol: LOC4344154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791180.1 | XP_015646666.1 | 13 - 14 | 2054 | 105 | |
| GenBank mRNA | AK072927.1 | - | 13 - 14 | 1978 | 105 | |
| AK100264.1 | - | 1 - 2 | 295 | 105 | ||
| AK100911.1 | - | 13 - 14 | 1918 | 105 | ||
| AK104043.1 | - | 1 - 2 | 104 | 105 | ||
More Information
Set Membership: |
|

LOC4344154
Genomic Map