Assay Details
| Gene Symbol: LOC4344154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791180.1 | XP_015646666.1 | 14 - 15 | 2146 | 69 | |
| GenBank mRNA | AK072927.1 | - | 14 - 15 | 2070 | 69 | |
| AK100264.1 | - | 2 - 3 | 387 | 69 | ||
| AK100911.1 | - | 14 - 15 | 2010 | 69 | ||
| AK104043.1 | - | 2 - 3 | 196 | 69 | ||
More Information
Set Membership: |
|

LOC4344154
Genomic Map