Assay Details
| Gene Symbol: LOC4344154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791180.1 | XP_015646666.1 | 18 - 19 | 2804 | 91 | |
| GenBank mRNA | AK072927.1 | - | 18 - 19 | 2728 | 91 | |
| AK100264.1 | - | 6 - 7 | 1045 | 91 | ||
| AK100911.1 | - | 18 - 19 | 2668 | 91 | ||
| AK104043.1 | - | 6 - 7 | 854 | 91 | ||
More Information
Set Membership: |
|

LOC4344154
Genomic Map