Assay Details
| Gene Symbol: LOC4344154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791180.1 | XP_015646666.1 | 4 - 5 | 811 | 117 | |
| GenBank mRNA | AK072927.1 | - | 4 - 5 | 735 | 117 | |
More Information
Set Membership: |
|

LOC4344154
Genomic Map