Assay Details
| Gene Symbol: LOC4344467 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795585.2 | XP_015651071.1 | 4 - 5 | 524 | 87 | |
| GenBank mRNA | AK067345.1 | - | 4 - 5 | 523 | 87 | |
| AK107316.1 | - | 4 - 5 | 525 | 87 | ||
More Information
Set Membership: |
|

LOC4344467
Genomic Map