Assay Details
| Gene Symbol: LOC4344467 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795585.2 | XP_015651071.1 | 5 - 6 | 640 | 100 | |
| GenBank mRNA | AK067345.1 | - | 5 - 6 | 639 | 100 | |
| AK107316.1 | - | 5 - 6 | 641 | 100 | ||
More Information
Set Membership: |
|

LOC4344467
Genomic Map