Assay Details
Gene Symbol: LOC4344938 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794915.1 | XP_015650401.1 | 6 - 7 | 1146 | 73 | |
GenBank mRNA | AK073315.1 | - | 6 - 7 | 1152 | 73 | |
AK120546.1 | - | 6 - 7 | 1152 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
