Assay Details
Gene Symbol: LOC4345201 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795636.1 | XP_015651122.1 | 1 - 2 | 513 | 101 | |
GenBank mRNA | AF456246.1 | - | 1 - 2 | 374 | 101 | |
AK066438.1 | - | 1 - 2 | 521 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
