Assay Details
Gene Symbol: LOC4345725 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794444.2 | XP_015649930.1 | 8 - 9 | 3866 | 82 | |
GenBank mRNA | AK102479.1 | - | 3 - 4 | 406 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
