Assay Details
| Gene Symbol: LOC4345872 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794667.1 | XP_015650153.1 | 1 - 2 | 495 | 72 | |
| GenBank mRNA | AK070359.1 | - | 1 - 2 | 450 | 72 | |
More Information
Set Membership: |
|

LOC4345872
Genomic Map