Assay Details
| Gene Symbol: LOC4345872 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794667.1 | XP_015650153.1 | 4 - 5 | 1294 | 67 | |
| GenBank mRNA | AK070359.1 | - | 4 - 5 | 1249 | 67 | |
| AK106196.1 | - | 3 - 4 | 690 | 67 | ||
More Information
Set Membership: |
|

LOC4345872
Genomic Map