Assay Details
| Gene Symbol: LOC4345911 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795481.2 | XP_015650967.1 | 5 - 6 | 2879 | 83 | |
| XM_015795482.2 | XP_015650968.1 | 5 - 6 | 2160 | 83 | ||
| GenBank mRNA | AK241274.1 | - | 5 - 6 | 2158 | 83 | |
More Information
Set Membership: |
|

LOC4345911
Genomic Map