Assay Details
| Gene Symbol: LOC4346340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792768.2 | XP_015648254.1 | 1 - 2 | 149 | 146 | |
| GenBank mRNA | AK062315.1 | - | 1 - 2 | 117 | 146 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015792768.2 transcripts.
Set Membership: |
|

LOC4346340
Genomic Map