Assay Details
Gene Symbol: LOC4347423 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795781.2 | XP_015651267.1 | 1 | 843 | 116 | |
GenBank mRNA | AK061067.1 | - | 1 | 758 | 116 | |
AK070140.1 | - | 1 | 759 | 116 | ||
AK106128.1 | - | 1 | 754 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
