Assay Details
| Gene Symbol: LOC9269832 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757203.2 | XP_015612689.1 | 9 - 10 | 1339 | 68 | |
| GenBank mRNA | AK060173.1 | - | 5 - 6 | 530 | 68 | |
| AK120151.1 | - | 9 - 10 | 1271 | 68 | ||
More Information
Set Membership: |
|

LOC9269832
Genomic Map