Assay Details
| Gene Symbol: LOC4347638 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755237.2 | XP_015610723.1 | 5 - 6 | 922 | 65 | |
| GenBank mRNA | AB097460.1 | - | 5 - 6 | 794 | 65 | |
| AK073610.1 | - | 5 - 6 | 866 | 65 | ||
| AK104977.1 | - | 5 - 6 | 810 | 65 | ||
More Information
Set Membership: |
|

LOC4347638
Genomic Map